Capillary columns, volatile, mineral oil
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7491 CP7491I5 CP749103 CP749106
VARICP7491EA 600 EUR
VARICP7491 VARICP7491I5 VARICP749103 VARICP749106
Capillary columns, volatile, mineral oil
Kolommen voor chromatografie
Select mineral oil provides the shortest analysis time is required with a stabilized, non-polar bonded phase specifically for fast mineral oil analysis.
  • Optimised selectivity for more reliable results
  • Low bleed for better accuracy
  • Available in UltiMetal for ultimate longevity
  • Available in economical three- and six-packs