Capillary columns, CP-Chirasil Val
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7494 CP7495 CP7495I5
VARICP7494EA 1620 EUR
VARICP7494 VARICP7495 VARICP7495I5
Capillary columns, CP-Chirasil Val
Kolommen voor chromatografie
The CP-Chirasil Val columns are designed for the separation of optically active compounds, especially amino acids. They have lower bleed levels than other phases with a Tmax of 200 °C, isothermally and programmed.
  • Both antipode phases available (D and L) for maximum versatility
  • Stabilized phase, over 50% cross-linked for great longevity
  • Specially designed and tested for amino acid enantiomers for the ultimate in reliable data