Drum funnels
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7740102 7740103 7740101
HIRS7740102EA 98 EUR
HIRS7740102 HIRS7740103 HIRS7740101
Drum funnels
Trechters Filtertrechters Trechters voor vaten
These large transparent funnels, made of PP, are suitable for filling large amounts of liquid.
  • Fast discharge by steep 60° angle
  • Convenient handle for hanging
  • Good chemical and temperature resistance