Safety shoes, lace-up, Jalcricket
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0247/42 J0247/45 J0247/41 J0247/43 J0247/44 J0247/36 J0247/37 J0247/38 J0247/39 J0247/40 J0247/46 J0247/47
JALCJ0247/42EA 97.6 EUR
JALCJ0247/42 JALCJ0247/45 JALCJ0247/41 JALCJ0247/43 JALCJ0247/44 JALCJ0247/36 JALCJ0247/37 JALCJ0247/38 JALCJ0247/39 JALCJ0247/40 JALCJ0247/46 JALCJ0247/47
Safety shoes, lace-up, Jalcricket
Schoenen en laarzen
Laced safety shoes made from perforated suede leather, 3D lining and Softane™/Néotril™ sole (heat resistant up to 300 °C); with padded tongue.
  • Lining with three dimensional foam structures for better ventilation
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Slip resistant Triftane™ sole helps to prevent fatigue
  • Puncture resistant stainless steel midsole
  • Perforated steel toecap (200 J)
Colour: Brown/sand
Certificaten: EN 345-1; S1P, HRO
Order Now
Specificaties
Environmentally Preferable