Safety ankle boots, lace-up, Jalmont
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0251/44 J0251/37 J0251/38 J0251/40 J0251/42 J0251/43 J0251/45 J0251/46 J0251/47 J0251/48 J0251/36 J0251/39 J0251/41
JALCJ0251/44EA 0 EUR
JALCJ0251/44 JALCJ0251/37 JALCJ0251/38 JALCJ0251/40 JALCJ0251/42 JALCJ0251/43 JALCJ0251/45 JALCJ0251/46 JALCJ0251/47 JALCJ0251/48 JALCJ0251/36 JALCJ0251/39 JALCJ0251/41
Safety ankle boots, lace-up, Jalmont
Schoenen en laarzen
Ankle boots made from full grain water repellent leather upper, 3D lining and Softane™ dual-density PU sole (resistant to abrasion, fuels, oils and grease); with padded tongue and ergonomic collar.
  • Lining with three dimensional foam structures for excellent ventilation
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Aérfort perforated steel toecap (200 J)
  • Puncture resistant stainless steel midsole
Colour: Dark red
Certificaten: EN 345-1; S3