Safety boots, pull-on, Jalartic
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0257/43 J0257/40 J0257/46 J0257/44 J0257/45 J0257/41 J0257/39 J0257/42 J0257/47 J0257/48 J0257/37 J0257/38 J0257/36
JALCJ0257/43EA 168 EUR
JALCJ0257/43 JALCJ0257/40 JALCJ0257/46 JALCJ0257/44 JALCJ0257/45 JALCJ0257/41 JALCJ0257/39 JALCJ0257/42 JALCJ0257/47 JALCJ0257/48 JALCJ0257/37 JALCJ0257/38 JALCJ0257/36
Safety boots, pull-on, Jalartic
Schoenen en laarzen
Pull-on winter boots made from AP23™ leather (oil and water repellent). It is 10 to 15% thicker than most leathers used for safety footwear (2,4 mm).
  • Warm fur for isothermal protection
  • Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Perforated steel toecap (200 J)
  • Puncture resistant stainless steel midsole
Colour: Honey
Certificaten: EN 345-1; S3, CI