Capillary columns, CP-Cyclodextrin-β-2,3,6-M-19
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7501 CP7500 CP7500I5
561-7501EA 1950 EUR
561-7501 VARICP7500 VARICP7500I5
Capillary columns, CP-Cyclodextrin-β-2,3,6-M-19
Kolommen voor chromatografie
The CP-Cyclodextrin-β-2,3,6-M-19 column separates many optical isomers that could not be analyzed previously.
  • Unique selectivity for isomer separation with ease-of-use
  • High inertness delivers accurate results
  • High efficiency for a broad application area
  • Useful for non-chiral compounds
CP-Cyclodextrin-β-2,3,6-M-19 has a very high inertness, enabling separation of underivatized polar compounds.