Capillary columns, CP-TCEP for alcohols in gasoline
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7525 CP7525I5
VARICP7525EA 1690 EUR
VARICP7525 VARICP7525I5
Capillary columns, CP-TCEP for alcohols in gasoline
Kolommen voor chromatografie
CP-TCEP column is able to separate benzene after n-dodecane. In addition, the excellent separation power provides the necessary resolution for complex mixtures such as gasoline.
  • Guaranteed analysis of alcohols in gasoline
  • Perfect peak shape for accurate separations of alcohols
  • High temperature stability to 135 °C for high productivity and enhanced longevity