Solid phase extraction cartridges, Bond Elut™ Sodium Sulphate Drying
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12162051B 12162052B 12162054B 12131033 12132004
VARI12162051BEA 389 EUR
VARI12162051B VARI12162052B VARI12162054B VARI12131033 VARI12132004
Solid phase extraction cartridges, Bond Elut™ Sodium Sulphate Drying
Kolommen voor chromatografie
Bond Elut™ Jr cartridges have top and bottom Luer fittings allowing easy sample processing when used in conjunction with standard SPE cartridges. Bond Elut™ LRC cartridges have a large reservoir above the sorbent bed and are suitable for use on any standard SPE vacuum manifold.
  • Highly effective pre-packed dessicant
  • Clean ACS grade, anhydrous sodium sulphate
  • Prepacked for convenience