Screw neck vials, N 9
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702282 702284 702283 702007 702008 702293 702006 702873 702009 702135 702010 702078 702079 702088 702172 702177 702178 702179 702335 702500 702857 702858 702863 702864 702874
MANA702282EA 15.9 EUR
MANA702282 MANA702284 MANA702283 MANA702007 MANA702008 MANA702293 MANA702006 MANA702873 MANA702009 MANA702135 MANA702010 MANA702078 MANA702079 MANA702088 MANA702172 MANA702177 MANA702178 MANA702179 MANA702335 MANA702500 MANA702857 MANA702858 MANA702863 MANA702864 MANA702874
Screw neck vials, N 9
flesjes
These screw neck vials are characterized by a wide opening and a short thread.
  • Made of clear and amber glass or transparent polypropylene
  • Silanised glass vials for highest demands
  • Available with flat, round and conical bottom or as micro-vials
  • Label and scale for convenient handling
  • Different inserts available
Can be used on almost all HPLC and GC autosamplers of Agilent, CTC, Dionex (Thermo Scientific), Knauer, PerkinElmer, Shimadzu, Thermo Scientific, Varian (Agilent), VWR® (Merck / Hitachi), Waters and others.
Polyproylene vials are suitable forMore Product Information