Robotic OMIX™ tips for Tomtec Quadra
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A57303 A57311
VARIA57303EA 0 EUR
VARIA57303 VARIA57311
Robotic OMIX™ tips for Tomtec Quadra
Pipettips
These Tomtec compatible tips contain a slice of monolithic SPE material.This allows for vacuum-free processing and walk-away automation. With hands-free SPE the process becomes much more streamlined and reproducible.
  • Fast, uniform flow maximises productivity and reproducibility
  • Small monolithic tip delivers low elution volumes, increasing sensitivity and reducing solvent usage
  • Vacuum-free processing improves reproducibility and shortens processing times