Crimp caps for crimp neck vials, N 8
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702800 70289 702025 702878
MANA702800EA 3.9 EUR
MANA702800 MANA70289 MANA702025 MANA702878
Crimp caps for crimp neck vials, N 8
Sluitingen Sluitingen voor chromatografievials
Ready-to-use crimp caps with assembled septa are available for the crimp neck vials.
  • High purity silicone/PTFE septa for more demanding analyses
  • Economic natural rubber/butyl/TEF or Red Rubber/FEP septa