Crimp caps for crimp neck vials, N 13
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70257 702802 70232 702803
MANA70257EA 62.1 EUR
MANA70257 MANA702802 MANA70232 MANA702803
Crimp caps for crimp neck vials, N 13
Sluitingen Sluitingen voor chromatografievials
The crimp caps with assembled septa are ready-to-use.
  • Standard cap with center hole or tamper-evident closure with center tear off cap
  • Molded septum with centrical PTFE lamination for very good sealing properties (typically called Pharma-Fix)