Snap cap vials, N 18 and N 22
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70271 70272 70273 702019
MANA70271EA 18.2 EUR
MANA70271 MANA70272 MANA70273 MANA702019
Snap cap vials, N 18 and N 22
flesjes
These snap cap vials are used for storage of powdery samples.
  • Made of clear glass (3rd hydrolytic class)
  • Flat bottom
Snap caps for both neck sizes are available.