Funnels, Stainless Steel
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A244-120 A244-50 A244-100
SAMPA244-120EA 69 EUR
SAMPA244-120 SAMPA244-50 SAMPA244-100
Funnels, Stainless Steel
Trechters Filtertrechters Filtertrechters
These stainless steel funnels are ideal for use with many type of liquids. Made from durable 304 grade stainless steel.
  • Crevice free construction
  • Fluted spout to aid product flow
  • Excellent chemical and temperature resistance