Bowls, disposable
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8017H-05 8017H-05S
216-1554EA 286 EUR
216-1554 SAMP8017H-05S
Bowls, disposable
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
HDPE, white.
  • Moulded and packed in a cleanroom
  • Individually bagged
  • BSE- and TSE-free
  • Full batch traceability
Ideal for handling and mixing powders and liquids. With spout and grip.
Certificaten: FDA acceptable CFR 177.1520, EU Regulations 10/2011 compliant