Safety helmets, Concept Roofer
Leverancier: CENTURION SAFETY PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S09EGRJ S09CYRF S09BRJ S09GRJ S09WRJ S09YRJ
111-0864EA 0 EUR
111-0864 111-0870 111-1308 111-1522 111-1313 111-1315
Safety helmets, Concept Roofer
Hoofdbescherming Veiligheidshelmen
The Concept Roofer is a light, strong and comfortable helmet made from ABS.
  • With premium ratchet headband ensuring secure fit
  • Six point terylene webbing cradle
  • Hydro-flock sweatband
Size: 520 to 630 mm
Weight: 340 g
EN 397
Certificaten: EN 397