Disposable earplugs, Contours®
Leverancier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7400 7403 7450 7453
111-1166EA 55.3 EUR
111-1166 111-1167 111-1169 111-1170
Disposable earplugs, Contours®
Oordoppen Oordoppen voor eenmalig gebruik
These extremely comfortable earplugs made of soft polyurethane foam are available in two different sizes.
  • Anatomical shape easily adapts to the ear canal: Less pressure and more comfort
  • Large base helps provide proper insertion, fit, and removal
  • Bright colour for improved visibility
SNR: 35 dB
Certificaten: EN 352-2:2002
Leveringsinformatie: Supplied in a box or in the hygienic dispenser system MoldexStation.
Verpakking: In pairs.