Screw caps for screw neck vials, N 10
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702046 702049 702044 702048 702047
MANA702046EA 33.4 EUR
MANA702046 MANA702049 MANA702044 MANA702048 MANA702047
Screw caps for screw neck vials, N 10
Sluitingen Sluitingen voor chromatografievials
Screw closures are bonded (septa firmly connected with the cap) or ready assembled.
  • Large range of bonded screw caps with different septum materials
  • Septa made from economical Red Rubber/TEF or high quality silicone/PTFE (also slitted)