Safety boots, lace-up, Jalarcher
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JJV28/43 JJV28/45 JJV28/38 JJV28/37 JJV28/39 JJV28/40 JJV28/41 JJV28/42 JJV28/44 JJV28/46 JJV28/47
JALCJJV28/43EA 167 EUR
JALCJJV28/43 JALCJJV28/45 JALCJJV28/38 JALCJJV28/37 JALCJJV28/39 JALCJJV28/40 JALCJJV28/41 JALCJJV28/42 JALCJJV28/44 JALCJJV28/46 JALCJJV28/47
Safety boots, lace-up, Jalarcher
Schoenen en laarzen
Metal-free boots made from full grain water repellent leather upper, 3D lining and Kompact Vibram® sole (fuil and oil resistant; contact heat resistant up to 300 °C HRO); with padded tongue, ergonomic collar and anti-abrasion PU scuff cap.
  • Lining with three dimensional foam structures for excellent ventilation
  • Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
  • Compact Vibram® sole with superb adherence and grip, high anti-abrasion value and 6,5 mm high self-cleaning cleats
  • Anti-abrasion heel protection and reflecting insert for improved visibility
  • YKK water proof zip for quick release with velcro protection
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Colour: Black
Certificaten: EN ISO 20345:2007; S3, HRO, SRC