EPA screw neck vials, N 24
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702022 702021 702024 702023 702133 702132 702131 702074 702831 702865 702877 702884
MANA702022EA 53.5 EUR
MANA702022 MANA702021 MANA702024 MANA702023 MANA702133 MANA702132 MANA702131 MANA702074 MANA702831 MANA702865 MANA702877 MANA702884
EPA screw neck vials, N 24
flesjes
These screw neck vials are preferential used for VOC and TOC analyses.
  • Made of clear and amber glass
  • Available with flat and round bottom
Can be used on autosamplers for VOC and TOC analyses of Agilent (AQUATek 70, SOLATek 72, Archon Purge+Trap), Dionex (Thermo Scientific - ASE 200, ASE 350, AS-HV), PerkinElmer (AQUATek 100), Shimadzu (ASI-L, ASI-V), Thermo Scientific (HiPerTOC), and Varian (Agilent - Archon).
With closed top of screw closures the vials are used for sample storage.
Bonded and ready assembled screw closures are available forMore Product Information