Softshell jacket, Pulco 622Z S.E.P.P.
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
622Z/B68/XS 622Z/B68/S 622Z/B68/M 622Z/B68/L 622Z/B68/1XL 622Z/B68/2XL 622Z/B68/3XL 622Z/M44/XS 622Z/M44/S 622Z/M44/M 622Z/M44/L 622Z/M44/1XL 622Z/M44/2XL 622Z/M44/3XL 622Z/N15/XS 622Z/N15/S 622Z/N15/M 622Z/N15/L 622Z/N15/1XL 622Z/N15/2XL 622Z/N15/3XL
113-07477EA 82.3 EUR
113-07477 113-07478 113-07479 113-07480 113-07481 113-07482 113-07483 113-07484 113-07485 113-07486 113-07487 113-07488 113-07489 113-07490 113-07491 113-07492 113-07493 113-07494 113-07495 113-07496 113-07497
Softshell jacket, Pulco 622Z S.E.P.P.
Werkkleding
Modern and lightweight outdoor jacket made of 2-layer bonded softshell (100% polyester stretch + 100% polyester fleece inside) with stitched seams. Provides good protection against cold temperatures and wind. The jacket can be zipped into the rain jacket 608Z for even better protection.
  • Straight collar, zip closure with chin protection and extra long zip pulls
  • One inset chest pocket and two inset front pockets with zip closure
  • Round inset sleeves, sleeve narrowing by touch and close fastening
Back length: 75 cm
Weight: 250 g/m²
EN 14058 (1XXXX)
Certificaten: EN 14058 (1XXXX)