PCR Plates for thermal cyclers
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPL0240/50 SPL0241/50 SPL0960/50 SPL0961/50
ADVASPL0240/50EA 123 EUR
ADVASPL0240/50 ADVASPL0241/50 ADVASPL0960/50 ADVASPL0961/50
PCR Plates for thermal cyclers
Microplaten PCR-platen
Piko® PCR plates utilise ultra-thin wall (UTW) technology to dramatically reduce the thickness of the well walls, and are specifically designed for use in the Piko® thermal cycler and the PikoReal® Real Time PCR system. Piko® PCR plates have the footprint of a microscope slide. Four Piko® 24-well (96-well) plates can be snapped into the plate frame to create a standard 96-well (384-well) plate.
  • Ultra-thin wall (UTW) for fast PCR/qPCR applications
  • Low profile
  • Compatible with standard multi-channel pipettes and liquid handling platforms
  • Well spacing and footprint conform to industry dimensions
These plates are made of PP.