SPE columns, HyperSep™ Verify™-CX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60108-769 60108-722 60108-721 60108-720 60108-742 60108-719 60108-723 60108-724 60108-741 60108-777
549-1061EA 220 EUR
549-1061 549-1063 HYPE60108-721 HYPE60108-720 549-1064 HYPE60108-719 HYPE60108-723 HYPE60108-724 HYPE60108-741 549-1062
SPE columns, HyperSep™ Verify™-CX
Kolommen voor chromatografie
Recommended for analysis of a wide range of drugs of abuse from biological matrices, including basic and neutral drugs.
  • Mixed mode sorbent for extraction of basic drugs
  • Separation based on two functional groups: reversed phase C8 and an ion exchanger, benzene sulfonic acid
  • Co-polymerized on a rigid, purified silica gel support