SPE columns, HyperSep™ Verify™-AX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60108-728 60108-764 60108-727 60108-730 60108-731 60108-732 60108-767
HYPE60108-728EA 238 EUR
HYPE60108-728 549-1066 HYPE60108-727 549-1065 549-1067 HYPE60108-732 HYPE60108-767
SPE columns, HyperSep™ Verify™-AX
Kolommen voor chromatografie
Features non-polar and cationic characteristics for improved analysis of acidic drugs and metabolites.
  • Mixed mode sorbent suited for the extraction of acidic drugs and metabolites from biological matrices
  • Separation based on two functional groups: reversed phase C8 and an ion exchanger, quaternary amine
  • Rigid, purified silica gel support to which the two functionalities are co-polymerised