Solid phase extraction cartridges, HyperSep™ SCX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60108-520 60108-433 60108-423 60108-716 60108-717 60108-718 60108-857 60108-420 60108-421 60108-422
HYPE60108-520EA 284 EUR
HYPE60108-520 HYPE60108-433 HYPE60108-423 HYPE60108-716 HYPE60108-717 HYPE60108-718 HYPE60108-857 HYPE60108-420 HYPE60108-421 HYPE60108-422
Solid phase extraction cartridges, HyperSep™ SCX
Kolommen voor chromatografie
A strong cation exchange sorbent for extraction of charged basic compounds.
  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent