Accessories for vacuum manifolds, CHROMABOND®
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
730152 730106 730157 730173 730174 730175 730176 730178 730189.1 730189.12 730233 730243 730366 730386 730387 730183.12 730184.12 730194 730990
MANA730152EA 90.6 EUR
MANA730152 MANA730106 MANA730157 MANA730173 MANA730174 MANA730175 MANA730176 MANA730178 MANA730189.1 MANA730189.12 MANA730233 MANA730243 MANA730366 MANA730386 MANA730387 MANA730183.12 MANA730184.12 MANA730194 MANA730990
Accessories for vacuum manifolds, CHROMABOND®
Spruitstukken
For the CHROMABOND® vacuum manifolds for upto 12, 16 or 24 samples the following accessories are available.
  • Replacement parts
  • Drying attachment for evaporation of eluates
  • Products for protection from cross contamination
  • Tubing adapters for application of large sample volumes