Digestion tubes, plain neck
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TT100/26 TT250/42
201-3050EA 152 EUR
201-3050 201-3051
Digestion tubes, plain neck
Buisjes Destructiebuizen
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Designed for use with block heaters.
Order Now
Specificaties
Environmentally Preferable