Tabard for Ladies, S843
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S843NARXXL S843NARS/M S843NARL/XL S843WHRL/XL S843WHRS/M S843WHRXXL S843RERL/XL S843RERS/M S843RERXXL
113-7589EA 14.2 EUR
113-7589 113-7588 113-7587 PRTWS843WHRL/XL PRTWS843WHRS/M PRTWS843WHRXXL PRTWS843RERL/XL PRTWS843RERS/M PRTWS843RERXXL
Tabard for Ladies, S843
Werkkleding
This tabard, made of Kingsmill fabric (65% polyester, 35% cotton), is ideal for use in the food industry.
  • Self-coloured binding
  • Adjustable side straps
  • Front pocket
Length: 790 mm (size S/M), 860 mm (sizes L/XL/XXL)
Weight: 245 g/m²