Overalls with bib and braces, Portwest Texo, TX12
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TX12BKRS TX12NARXXL TX12NARXL TX12NARS TX12NARM TX12NARL TX12GRRXXXL TX12GRRXXL TX12GRRXL TX12GRRS TX12GRRM TX12GRRL TX12BKRXXXL TX12BKRXL TX12BKRM TX12BKRL TX12BKRXXL TX12NARXXXL
113-9687EA 41.8 EUR
113-9687 113-9698 113-9697 113-9696 113-9695 113-9694 113-9693 113-9692 113-9691 113-7636 113-7635 113-7634 113-9690 113-9688 113-9686 113-9685 113-9689 113-9699
Overalls with bib and braces, Portwest Texo, TX12
Werkkleding
These overalls made of Portwest Texo fabric (60% cotton, 40% polyester), provide complete lower body and leg protection. The cotton ensures the trousers wash well at 60 °C while the polyester gives a ruggedness essential for daily use.
  • Back elasticated waist for improved comfort
  • Kneepad pockets, mobile phone pocket, thigh pocket, concealed bib pocket
  • Velcro® at hems for easy adjustment
  • Compatible with the Contrast Jacket TX10
Weight: 245 g/m²