Capillary columns, PERMABOND® CW 20 M-DEG
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
723063.25 723327.25
MANA723063.25EA 728 EUR
MANA723063.25 MANA723327.25
Capillary columns, PERMABOND® CW 20 M-DEG
Kolommen voor chromatografie
Special GC phase for the analysis of diethylene glycol.
  • Polar phase, tested for diethylene glycol determination
  • Max. temperature for isothermal operation 220 °C
  • Max. temperature for short isotherms in a temperature program 240 °C
USP code: G16.