Safety padlocks, with steel shackle
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
051346 051339 051344 813638 813636 813637 051345 051347 051353 814113
238-0029EA 123.22 EUR
238-0029 238-0026 238-0027 238-0050 238-0048 238-0049 238-0028 238-0030 238-0031 238-0081
Safety padlocks, with steel shackle
Veiligheidssloten
These compact and lightweight padlocks are ideal for use with all lockout equipment. The insulated key chamber protects workers from shocks when the key is inserted - ideal for all electrical applications, especially in process industries.
  • Reinforced nylon, with hardened steel shackle (38 mm)
  • Key will not be released until the padlock shackle is closed
  • 6-pin locking mechanism cylinder (rugged nylon) is ribbed in design for easy handling and grip
  • Nylon casing is chemical and corrosion resistant with enhanced impact resistance
  • Each padlock is keyed differently and is supplied with one key
Shackle diameter: 6,5 mm