Hand cleansing cream, Solopol® Classic EXTRA/Solopol® strong
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35577 SCU2LT
115-1505EA 155.88 EUR
115-1505 115-2348
Hand cleansing cream, Solopol® Classic EXTRA/Solopol® strong
Huidverzorging Zepen en huidreinigingsmiddelen
Solvent-free hand cleanser paste with scrubbing agent for extra heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in industrial environments.
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • Contains Eucornol® skin protectant to minimise the risk of skin irritation
  • Petroleum solvent-free, silicone-free, colourant-free, perfumed
Certificaten: EU Ecolabel
Bestelinformatie: Soft bottle may only be used with Stoko®More Product Information