Polo shirts, long sleeved, B212
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B212NARXXL B212NARXL B212NARS B212NARM B212NARL B212BKRS B212BKRXS B212BKRL B212BKRM B212BKRXXXL B212BKRXXL B212BKRXL
113-6940EA 18.7 EUR
113-6940 113-6939 113-6938 113-6937 113-6936 PRTWB212BKRS PRTWB212BKRXS PRTWB212BKRL PRTWB212BKRM PRTWB212BKRXXXL PRTWB212BKRXXL PRTWB212BKRXL
Polo shirts, long sleeved, B212
Werkkleding
This practical long sleeved polo shirt offers full upper body comfort for casual wear. Manufactured from 65% polyester and 35% cotton.
  • Rib knitted collar and cuffs
  • Three button placket and matching buttons
  • Heavyweight polyester and cotton mix ensures garment durability
Weight: 210 g/m²