Cholesterol pentyl carbonate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C0720-1G C0720-10G
TCIAC0720-1GEA 41.8 EUR
TCIAC0720-1G TCIAC0720-10G
Cholesterol pentyl carbonate
Cholesterol pentyl carbonate
Formule: C₃₃H₅₆O₃
MDL Nummer: MFCD00059503
CAS nummer: 15455-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30cd6a4e79eeb7f57f5c9a183864bd14ead8d614&vid=30cd6a4e79eeb7f57f5c9a183864bd14ead8d614&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Melting point 103.0 to 107.0 deg-C
Specific rotation [a]20/D -30.0 to -39.0 deg (C=2,CHCl3)
Solubility in Chloroform almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR