Allyl phenyl carbonate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Allyl Phenyl Carbonate, ['Carbonic acid Allyl Phenyl Ester']
A2303-25G
A2303-5G
TCIAA2303-25GEA
209
EUR
TCIAA2303-25G
TCIAA2303-5G
Allyl phenyl carbonate ≥97.0% (by GC)
Allyl phenyl carbonate
Formule:
C₁₀H₁₀O₃ Kookpunt: 78°C |
MDL Nummer:
MFCD07784342 CAS nummer: 16308-68-2 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...