Methyl propyl disulphide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M1781-25G M1781-5G
TCIAM1781-25GEA 136 EUR
TCIAM1781-25G TCIAM1781-5G
Methyl propyl disulphide ≥97.0% (by GC)
Methyl propyl disulphide
Formule: C₄H₁₀S₂
Kookpunt: 71°C
Vlampunt: 43°C
MDL Nummer: MFCD00013400
CAS nummer: 2179-60-4
EINECS: 218-551-2
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf1627c9137d8a6190595d6dde52e2b419a78428&vid=cf1627c9137d8a6190595d6dde52e2b419a78428&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR