4-Carboxyphenylboronic acid 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
344400010 344400050 344400250
ACRO344400010EA 36.4 EUR
ACRO344400010 ACRO344400050 ACRO344400250
4-Carboxyphenylboronic acid 97%
4-Carboxyphenylboronic acid
Formule: C₇H₇BO₄
Smeltpunt: 238…240 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151801
CAS nummer: 14047-29-1
Order Now
Specification Test Results
Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
Specificaties
Environmentally Preferable