3-Iodobenzylamine hydrochloride ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0710-1G I0710-5G
TCIAI0710-1GEA 22 EUR
TCIAI0710-1G TCIAI0710-5G
3-Iodobenzylamine hydrochloride ≥98.0% (by HPLC)
3-Iodobenzylamine hydrochloride
Formule: C₇H₉ClIN
Smeltpunt: 194°C
MDL Nummer: MFCD00012857
CAS nummer: 3718-88-5
EINECS: 223-068-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42b226e25dc2b9b09a03eba1fdf38d26da60393c&vid=42b226e25dc2b9b09a03eba1fdf38d26da60393c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR