2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A0397-1G A0397-25G A0397-5G
TCIAA0397-1GEA 38.5 EUR
TCIAA0397-1G TCIAA0397-25G TCIAA0397-5G
2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)
2-Amino-4-phenylphenol
Formule: C₁₂H₁₁NO
MDL Nummer: MFCD00059187
CAS nummer: 1134-36-7
EINECS: 214-484-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9980011df2f29025c40bad77e66417b07a227a32&vid=9980011df2f29025c40bad77e66417b07a227a32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR