Cyclohexyl methyl sulphide ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
L03712.06
L03712.14
L03712.06EA
46.6
EUR
L03712.06
L03712.14
Cyclohexyl methyl sulphide ≥97%
Cyclohexyl methyl sulphide
Purity: 97%
CAS No.: 7133-37-1
Molecular Formula: C7H14S
Formula Weight: 130.25
Flash Point: 61°(142°F)
Refractive Index: 1.4940
Odour: Unpleasant
Melting Point: -80°
Boiling Point: 89-90°/35mm
Beilstein Reference: 1919309
UN No.: 3334
MDL No.: MFCD00039460
Formule:
C₇H₁₄S MW: 130.25 g/mol Kookpunt: 89…90 °C (35 mmHg) Smeltpunt: –80 °C Dichtheid: 0.944 Vlampunt: 61 °C (142 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00039460 CAS nummer: 7133-37-1 EINECS: 230-431-1 ADR: 9 III |