Sodium-1-pentadecanesulphonate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0889-1G P0889-5G
TCIAP0889-1GEA 176 EUR
TCIAP0889-1G TCIAP0889-5G
Sodium-1-pentadecanesulphonate
Sodium-1-pentadecanesulphonate
Formule: C₁₅H₃₁NaO₃S
MDL Nummer: MFCD00059838
CAS nummer: 5896-54-8
EINECS: 227-588-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f953cebe033f8ee91f06f1c3e7cf0ecff9e2a3e0&vid=f953cebe033f8ee91f06f1c3e7cf0ecff9e2a3e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR