Sodium-1-tetradecanesulphonate

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T0757-1G T0757-10G
TCIAT0757-1GEA 26.4 EUR
TCIAT0757-1G TCIAT0757-10G
Sodium-1-tetradecanesulphonate
Sodium-1-tetradecanesulphonate
Formule: C₁₄H₂₉NaO₃S
MDL Nummer: MFCD00043415
CAS nummer: 6994-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87eb4a028b61233297fe1ff9b6d07fc24b5d36d4&vid=87eb4a028b61233297fe1ff9b6d07fc24b5d36d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Solubility in hot Water almost transparency
Drying loss max. 1.0 %
Ignition residue (Sulfate) 23.2 to 23.8 % (after drying)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR