1,3-Diisopropenylbenzene ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-tert-butylpyrocatechol (TBC)

D1957-25ML D1957-500ML
TCIAD1957-25MLEA 28.6 EUR
TCIAD1957-25ML TCIAD1957-500ML
1,3-Diisopropenylbenzene ≥97.0% (by GC) stabilised
1,3-Diisopropenylbenzene
Formule: C₁₂H₁₄
Kookpunt: 231°C
Vlampunt: 93°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008584
CAS nummer: 3748-13-8
EINECS: 223-146-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7d586b91af5611f72f831f8b1ccd9934c6bdbc0&vid=a7d586b91af5611f72f831f8b1ccd9934c6bdbc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR