1,10-Undecadiene ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

U0024-1ML U0024-5ML
TCIAU0024-1MLEA 105 EUR
TCIAU0024-1ML TCIAU0024-5ML
1,10-Undecadiene ≥90.0% (by GC)
1,10-Undecadiene
Formule: C₁₁H₂₀
Kookpunt: 187°C
MDL Nummer: MFCD00059203
CAS nummer: 13688-67-0
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75aa187a9bb08dc34581c34e9f6bf3492f796f94&vid=75aa187a9bb08dc34581c34e9f6bf3492f796f94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR