Methyl perfluorododecanoate ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1917-25G
M1917-5G
TCIAM1917-25GEA
162
EUR
TCIAM1917-25G
TCIAM1917-5G
Methyl perfluorododecanoate ≥95.0% (by GC)
Methyl perfluorododecanoate
Formule:
C₁₃H₃F₂₃O₂ |
MDL Nummer:
MFCD00236616 CAS nummer: 56554-52-0 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 54.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...