α,2-Dibromo-5-fluorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3784-25G B3784-5G
TCIAB3784-25GEA 79.2 EUR
TCIAB3784-25G TCIAB3784-5G
α,2-Dibromo-5-fluorotoluene ≥98.0% (by GC)
α,2-Dibromo-5-fluorotoluene
Formule: C₇H₅Br₂F
Kookpunt: 96°C
MDL Nummer: MFCD00042187
CAS nummer: 112399-50-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d87fd6adc58420f5d99c5d6731dfc1f0d0f6f204&vid=d87fd6adc58420f5d99c5d6731dfc1f0d0f6f204&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR