EDTA (ethylenediamine tetraacetic acid) ≥99.4% for electrophoresis
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
J65585.22
J65585.36
J65585.A4
J65585.22EA
31.6
EUR
J65585.22
J65585.36
J65585.A4
EDTA (ethylenediamine tetraacetic acid) ≥99.4% for electrophoresis
EDTA (ethylenediamine tetraacetic acid)
Purity: 99.4+%
CAS No.: 60-00-4
Molecular Formula: C10H16N2O8
Formula Weight: 292.24
Physical Form: Powder
Density: 0.68
Appearance: White
Melting Point: ca 245° dec.
Beilstein Reference: 1716295
Merck Reference: 14,3517
MDL No.: MFCD00003541
Formule:
C₁₀H₁₆N₂O₈ Kookpunt: 614 °C (1013 hPa) Dichtheid: 0,68 Vlampunt: >100 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003541 CAS nummer: 60-00-4 EINECS: 200-449-4 Bereik: 01-2119486399-18 Merck Index: 14,3517 |