Magnesium sulphate 1 M aqueous solution, Sterile-Filtered
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: MgSO4, Magnesium sulphate anhydrous
J61030.XE J61030.AK
J61030.XEEA 70 EUR
J61030.XE J61030.AK
Magnesium sulphate 1 M aqueous solution, Sterile-Filtered
Magnesium sulphate
CAS No.: 7487-88-9
Molecular Formula: MgO4S
Formula Weight: 120.37
Physical Form: Liquid
MDL No.: MFCD00011110
Formule: MgSO₄
MW: 120.37 g/mol
Smeltpunt: 1127
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011110
CAS nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM