Diindium trisulphate for synthesis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.43982.0100 8.43982.0025
8.43982.0100EA 424 EUR
8.43982.0100 8.43982.0025
Diindium trisulphate for synthesis
Diindium trisulphate
Formule: O₁₂In₂S₃
Smeltpunt: 585 °C
Dichtheid: 3,438 g/cm³ (20 °C)
MDL Nummer: MFCD00011059
CAS nummer: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978
Order Now
Specification Test Results
Molar Mass 517.83 g/mol